Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCS18
Entrez Gene ID
829603
Gene Name
3-ketoacyl-CoA synthase 18
Gene Aliases
3-ketoacyl-CoA synthase 18, AT4G34520, FAE1, FATTY ACID ELONGATION1, T4L20.100, T4L20_100
Chromosome Location
Chr.4: 16494205 - 16495842 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119617.2 NP_195178.1 1 - 1 1438 102

View Full Product Details