Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HBT
Entrez Gene ID
816519
Gene Name
CDC27 family protein
Gene Aliases
AT2G20000, CDC27b, HOBBIT, T2G17.20, T2G17_20
Chromosome Location
Chr.2: 8631959 - 8637300 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335657.1 NP_001323708.1 10 - 11 1767 124
NM_179663.3 NP_849994.1 10 - 11 1767 124
GenBank mRNA AJ487669.1 - 10 - 11 1767 124

View Full Product Details