Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RD2
Entrez Gene ID
816700
Gene Name
Adenine nucleotide alpha hydrolases-like superfamily protein
Gene Aliases
AT2G21620, F2G1.11, F2G1_11
Chromosome Location
Chr.2: 9248525 - 9250311 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179684.4 NP_850015.1 3 - 4 656 77
NM_179685.2 NP_850016.1 3 - 4 626 77
GenBank mRNA AB039925.1 - 3 - 4 487 77
AY087097.1 - 3 - 4 608 77

View Full Product Details