Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNGC14
Entrez Gene ID
816997
Gene Name
cyclic nucleotide-gated channel 14
Gene Aliases
AT2G24610, ATCNGC14, F25P17.9, F25P17_9, cyclic nucleotide-gated channel 14
Chromosome Location
Chr.2: 10456879 - 10460571 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335940.1 NP_001324934.1 2 - 3 1216 84
NM_179725.2 NP_850056.1 3 - 4 1006 84
GenBank mRNA AK228122.1 - 3 - 4 840 84

View Full Product Details