Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OVA4
Entrez Gene ID
817126
Gene Name
Nucleotidylyl transferase superfamily protein
Gene Aliases
AT2G25840, F17H15.13, F17H15_13, ovule abortion 4
Chromosome Location
Chr.2: 11021692 - 11025451 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336021.1 NP_001323723.1 6 - 7 796 116
NM_179739.4 NP_850070.1 6 - 7 952 116
NM_201805.1 NP_973534.1 6 - 7 703 116
GenBank mRNA AY081299.1 - 6 - 7 739 116

View Full Product Details