Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YAB5
Entrez Gene ID
817199
Gene Name
plant-specific transcription factor YABBY family protein
Gene Aliases
AT2G26580, T9J22.25, T9J22_25, YABBY5
Chromosome Location
Chr.2: 11303455 - 11307010 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179749.3 NP_850080.1 2 - 3 465 60
NM_179750.3 NP_850081.1 3 - 4 308 60
GenBank mRNA AK119091.1 - 2 - 3 234 60
BT003734.1 - 2 - 3 194 60

View Full Product Details