Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAT12
Entrez Gene ID
817328
Gene Name
nucleobase-ascorbate transporter 12
Gene Aliases
ARABIDOPSIS NUCLEOBASE-ASCORBATE TRANSPORTER 12, AT2G27810, ATNAT12, F15K20.9, F15K20_9, nucleobase-ascorbate transporter 12
Chromosome Location
Chr.2: 11852078 - 11856335 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084501.1 NP_001077970.1 2 - 3 889 99
NM_001336128.1 NP_001323574.1 2 - 3 889 99
NM_179777.2 NP_850108.1 2 - 3 891 99
NM_201821.2 NP_973550.2 2 - 3 889 99
GenBank mRNA AY099545.1 - 2 - 3 802 99
AY444865.1 - 2 - 3 631 99
BT002121.1 - 2 - 3 631 99

View Full Product Details