Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCRK
Entrez Gene ID
817371
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT2G28250, T3B23.8, T3B23_8
Chromosome Location
Chr.2: 12042348 - 12046616 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036358.2 NP_001031435.1 4 - 5 914 121
NM_001336153.1 NP_001323735.1 4 - 5 647 121
NM_001336154.1 NP_001323736.1 4 - 5 388 121
NM_001336155.1 NP_001323737.1 4 - 5 773 121
NM_001336156.1 NP_001323738.1 4 - 5 696 121
NM_179784.2 NP_850115.1 3 - 4 883 121
GenBank mRNA AY065457.1 - 3 - 4 435 121
BT004417.1 - 3 - 4 265 121

View Full Product Details