Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALB3
Entrez Gene ID
817429
Gene Name
63 kDa inner membrane family protein
Gene Aliases
ABL3, ALBINO 3, AT2G28800, CHLOROPLAST MEMBRANE PROTEIN ALB3, F8N16.9, F8N16_9
Chromosome Location
Chr.2: 12356321 - 12359568 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001124933.1 NP_001118405.1 2 - 3 579 65
NM_001202697.1 NP_001189626.1 2 - 3 578 65
NM_128439.3 NP_180446.1 2 - 3 895 65
NM_179794.2 NP_850125.1 2 - 3 579 65
GenBank mRNA BT002436.1 - 2 - 3 584 65
BT006602.1 - 2 - 3 485 65
U89272.1 - 2 - 3 521 65

View Full Product Details