Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMO2-2
Entrez Gene ID
817488
Gene Name
sterol 4-alpha-methyl-oxidase 2-2
Gene Aliases
AT2G29390, ATSMO2, ATSMO2-2, Arabidopsis thaliana sterol 4-alpha-methyl-oxidase 2-2, F16P2.23, F16P2_23, STEROL 4-ALPHA-METHYL-OXIDASE 2, sterol 4-alpha-methyl-oxidase 2-2
Chromosome Location
Chr.2: 12610508 - 12612937 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084505.1 NP_001077974.1 2 - 3 180 71
NM_001336210.1 NP_001318311.1 2 - 3 311 71
NM_128493.4 NP_565681.1 2 - 3 265 71
NM_179802.3 NP_850133.1 2 - 3 308 71
NM_201830.2 NP_973559.1 1 - 2 646 71
GenBank mRNA AF327853.1 - 2 - 3 170 71
AY086287.1 - 2 - 3 243 71
BT002780.1 - 2 - 3 235 71
BT005125.1 - 2 - 3 170 71

View Full Product Details