Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIFI
Entrez Gene ID
820827
Gene Name
post-illumination chlorophyll fluorescence increase
Gene Aliases
AT3G15840, post-illumination chlorophyll fluorescence increase
Chromosome Location
Chr.3: 5356587 - 5358596 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035629.1 NP_001030706.1 1 - 2 102 105
NM_001035630.1 NP_001030707.1 1 - 2 160 105
NM_001125170.1 NP_001118642.1 1 - 2 88 105
NM_112454.3 NP_566528.1 1 - 2 257 105
GenBank mRNA AY035025.1 - 1 - 2 116 105
AY133879.1 - 1 - 2 79 105

View Full Product Details