The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_112455.4 | NP_566529.1 | 1 - 2 | 1012 | 101 |
GenBank mRNA | AB113831.1 | - | 1 - 2 | 526 | 101 |
BT000957.1 | - | 1 - 2 | 526 | 101 |