Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAI2
Entrez Gene ID
820839
Gene Name
DNA topoisomerase-like protein
Gene Aliases
AT3G15950
Chromosome Location
Chr.3: 5397541 - 5402653 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035631.2 NP_001030708.1 2 - 3 175 103
NM_001338191.1 NP_001326809.1 2 - 3 175 103
NM_001338192.1 NP_001326807.1 2 - 3 175 103
NM_001338193.1 NP_001326808.1 2 - 3 176 103
NM_112465.4 NP_188216.2 2 - 3 175 103
GenBank mRNA AY120750.1 - 2 - 3 174 103
BT001207.1 - 2 - 3 133 103

View Full Product Details