Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAB6
Entrez Gene ID
820885
Gene Name
poly(A) binding protein 6
Gene Aliases
AT3G16380, T2O4.4, poly(A) binding protein 6
Chromosome Location
Chr.3: 5558445 - 5561126 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112509.2 NP_188259.1 4 - 5 1223 72
GenBank mRNA AX765882.1 - 4 - 5 1090 72

View Full Product Details