Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MPPalpha
- Entrez Gene ID
-
820896
- Gene Name
- mitochondrial processing peptidase alpha subunit
- Gene Aliases
-
AT3G16480, T2O4.13, mitochondrial processing peptidase alpha subunit
- Chromosome Location
-
Chr.3: 5599710 - 5603027 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112519.4 |
NP_566548.1 |
3 - 4 |
768 |
115 |
View Full Product Details