Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQD26
Entrez Gene ID
820897
Gene Name
IQ-domain 26
Gene Aliases
AT3G16490, IQ-domain 26
Chromosome Location
Chr.3: 5603848 - 5605765 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112520.4 NP_188270.1 1 - 2 642 115
GenBank mRNA BT015939.1 - 1 - 2 521 115
BT020577.1 - 1 - 2 366 115

View Full Product Details