Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGD15
Entrez Gene ID
821033
Gene Name
ARF-GAP domain 15
Gene Aliases
ARF-GAP domain 15, AT3G17660
Chromosome Location
Chr.3: 6037598 - 6039407 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338298.1 NP_001327025.1 4 - 5 376 81
NM_001338300.1 NP_001327026.1 4 - 5 376 81
NM_001338301.1 NP_001327027.1 3 - 4 263 81
NM_112647.4 NP_188393.2 4 - 5 376 81
GenBank mRNA AK228639.1 - 4 - 5 376 81

View Full Product Details