Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WOX1
Entrez Gene ID
821325
Gene Name
WUSCHEL related homeobox 1
Gene Aliases
AT3G18010, WUSCHEL related homeobox 1
Chromosome Location
Chr.3: 6160733 - 6163453 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112682.4 NP_188428.3 2 - 3 772 76

View Full Product Details