Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UNG
Entrez Gene ID
821394
Gene Name
uracil dna glycosylase
Gene Aliases
AT3G18630, ATUNG, uracil dna glycosylase
Chromosome Location
Chr.3: 6410958 - 6413196 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112749.5 NP_188493.1 4 - 5 903 72
GenBank mRNA AY084956.1 - 4 - 5 775 72
BT029175.1 - 4 - 5 709 72

View Full Product Details