Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VRN1
Entrez Gene ID
821432
Gene Name
AP2/B3-like transcriptional factor family protein
Gene Aliases
AT3G18990, REDUCED VERNALIZATION RESPONSE 1, REM39, REPRODUCTIVE MERISTEM 39
Chromosome Location
Chr.3: 6548127 - 6552055 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338374.1 NP_001326749.1 1 - 2 529 104
NM_112785.5 NP_188529.2 1 - 2 550 104
GenBank mRNA AB493620.1 - 1 - 2 63 104
AF289052.1 - 1 - 2 374 104
AK230388.1 - - 262 104
BT024731.1 - 1 - 2 63 104

View Full Product Details