Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG2
Entrez Gene ID
821453
Gene Name
autophagy 2
Gene Aliases
AT3G19190, ATATG2, AUTOPHAGY 2, PEROXISOME UNUSUAL POSITIONING 1, PEUP1
Chromosome Location
Chr.3: 6639639 - 6648870 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338385.1 NP_001327751.1 7 - 8 5088 135
NM_001338386.1 NP_001327752.1 7 - 8 5088 135
NM_112806.3 NP_188550.3 7 - 8 4836 135
GenBank mRNA HQ446214.1 - 7 - 8 4848 135

View Full Product Details