Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUB25
Entrez Gene ID
821472
Gene Name
plant U-box 25
Gene Aliases
AT3G19380, plant U-box 25
Chromosome Location
Chr.3: 6714133 - 6716178 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112825.4 NP_566632.1 1 - 1 1517 90
GenBank mRNA AY086503.1 - 1 - 1 1263 90
BT025982.1 - 1 - 1 1206 90

View Full Product Details