Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTL2
Entrez Gene ID
821587
Gene Name
RNAse THREE-like protein 2
Gene Aliases
AT3G20420, ATRTL2, RNASEIII-LIKE 2, RNAse THREE-like protein 2
Chromosome Location
Chr.3: 7119191 - 7121023 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112933.3 NP_566661.1 2 - 3 767 89

View Full Product Details