Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLGAMMA1
Entrez Gene ID
821600
Gene Name
polymerase gamma 1
Gene Aliases
AT3G20540, PolIB, polymerase I B, polymerase gamma 1
Chromosome Location
Chr.3: 7168054 - 7173617 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112946.5 NP_188690.3 1 - 2 322 64
GenBank mRNA AB211533.1 - 1 - 2 115 64
AK226811.1 - 1 - 2 248 64
AY195962.1 - 1 - 2 191 64

View Full Product Details