Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLGAMMA1
Entrez Gene ID
821600
Gene Name
polymerase gamma 1
Gene Aliases
AT3G20540, PolIB, polymerase I B, polymerase gamma 1
Chromosome Location
Chr.3: 7168054 - 7173617 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203013.1 NP_001189942.1 4 - 5 1092 126
NM_112946.5 NP_188690.3 3 - 4 1226 126
GenBank mRNA AB211533.1 - 3 - 4 1019 126
AK226811.1 - 3 - 4 1152 126
AY195962.1 - 3 - 4 1095 126

View Full Product Details