Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLGAMMA1
Entrez Gene ID
821600
Gene Name
polymerase gamma 1
Gene Aliases
AT3G20540, PolIB, polymerase I B, polymerase gamma 1
Chromosome Location
Chr.3: 7168054 - 7173617 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203013.1 NP_001189942.1 6 - 7 2242 86
NM_112946.5 NP_188690.3 5 - 6 2376 86
GenBank mRNA AB211533.1 - 5 - 6 2169 86
AK226811.1 - 5 - 6 2302 86
AY195962.1 - 5 - 6 2245 86

View Full Product Details