Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCB2;2
Entrez Gene ID
829714
Gene Name
Cyclin B2;2
Gene Aliases
AT4G35620, Cyclin B2;2, F8D20.130, F8D20_130
Chromosome Location
Chr.4: 16901596 - 16904001 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342363.1 NP_001329476.1 4 - 5 557 141
NM_119727.4 NP_195287.1 4 - 5 557 141
GenBank mRNA AK226721.1 - 4 - 5 539 141

View Full Product Details