Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GB2
Entrez Gene ID
829740
Gene Name
GTP-binding 2
Gene Aliases
AT4G35860, ATGB2, ATRAB2C, ATRABB1B, F4B14.130, F4B14_130, GTP-BINDING PROTEIN ATGB2, GTP-binding 2
Chromosome Location
Chr.4: 16986840 - 16989047 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085030.1 NP_001078499.1 2 - 3 590 63
NM_119752.4 NP_195311.1 3 - 4 476 63
GenBank mRNA AY065262.1 - 3 - 4 405 63
AY085750.1 - 3 - 4 405 63
AY117348.1 - 3 - 4 268 63
U46925.1 - 3 - 4 395 63

View Full Product Details