Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AP2
Entrez Gene ID
829845
Gene Name
Integrase-type DNA-binding superfamily protein
Gene Aliases
AP22.49, AP22_49, APETALA 2, AT4G36920, AtAP2, FL1, FLO2, FLORAL HOMEOTIC PROTEIN APETALA 2, FLORAL MUTANT 2, FLOWER 1
Chromosome Location
Chr.4: 17400610 - 17403469 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204009.1 NP_001190938.1 9 - 10 1094 117
NM_119856.3 NP_195410.1 8 - 9 1253 117
GenBank mRNA U12546.1 - 8 - 9 1168 117

View Full Product Details