Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP1D
Entrez Gene ID
829858
Gene Name
methionine aminopeptidase 1D
Gene Aliases
AT4G37040, methionine aminopeptidase 1D
Chromosome Location
Chr.4: 17455100 - 17457357 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119867.3 NP_568014.1 1 - 2 183 93
GenBank mRNA AF250963.1 - 1 - 2 144 93
AY085839.1 - 1 - 2 141 93

View Full Product Details