Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMA1
Entrez Gene ID
829881
Gene Name
heavy metal atpase 1
Gene Aliases
AP22.4, AP22_4, ARABIDOPSIS THALIANA HEAVY METAL ATPASE 1, AT4G37270, ATHMA1, heavy metal atpase 1
Chromosome Location
Chr.4: 17541771 - 17546546 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119890.7 NP_195444.1 2 - 3 851 88
GenBank mRNA AJ872069.1 - 2 - 3 683 88

View Full Product Details