Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGP18
Entrez Gene ID
829900
Gene Name
arabinogalactan protein 18
Gene Aliases
ARABINOGALACTAN PROTEIN AGP18, AT4G37450, ATAGP18, F6G17.100, F6G17_100, arabinogalactan protein 18, arabinogalactan protein 18
Chromosome Location
Chr.4: 17605538 - 17607050 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342442.1 NP_001329215.1 1 - 2 856 86
NM_119909.3 NP_568027.1 1 - 2 730 86
GenBank mRNA AF305940.1 - 1 - 2 607 86

View Full Product Details