Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRFR1
Entrez Gene ID
829901
Gene Name
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases
AT4G37460, F6G17.110, F6G17_110, SUPPRESSOR OF RPS4-RLD 1
Chromosome Location
Chr.4: 17608378 - 17615946 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342443.1 NP_001329700.1 4 - 5 1382 93
NM_119910.4 NP_195462.3 4 - 5 1382 93

View Full Product Details