Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCD5;1
Entrez Gene ID
829917
Gene Name
cyclin d5;1
Gene Aliases
AT4G37630, F19F18.120, F19F18_120, cyclin d5;1
Chromosome Location
Chr.4: 17679168 - 17681264 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342450.1 NP_001320153.1 1 - 2 392 87
NM_001342451.1 NP_001328589.1 1 - 2 577 87
NM_119926.5 NP_195478.2 1 - 2 577 87
GenBank mRNA AK221431.1 - 1 - 2 466 87
BT011732.1 - 1 - 2 248 87

View Full Product Details