Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mtHsc70-1
Entrez Gene ID
829947
Gene Name
mitochondrial heat shock protein 70-1
Gene Aliases
AT4G37910, F20D10.30, F20D10_30, mitochondrial heat shock protein 70-1
Chromosome Location
Chr.4: 17824777 - 17828192 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342461.1 NP_001328002.1 5 - 6 1491 79
NM_119952.4 NP_195504.2 5 - 6 1479 79
GenBank mRNA BT002390.1 - 5 - 6 1476 79
BT008430.1 - 5 - 6 1398 79

View Full Product Details