Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELI3-1
Entrez Gene ID
829954
Gene Name
cinnamyl alcohol dehydrogenase 7
Gene Aliases
AT4G37980, ATCAD7, CAD7, CINNAMYL-ALCOHOL DEHYDROGENASE 7, ELI3, ELICITOR-ACTIVATED GENE 3, F20D10.100, F20D10_100, elicitor-activated gene 3-1
Chromosome Location
Chr.4: 17852441 - 17854608 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036728.2 NP_001031805.1 2 - 3 430 104
NM_119959.4 NP_195511.1 2 - 3 430 104
GenBank mRNA AF360225.1 - 2 - 3 250 104
AK317050.1 - 2 - 3 250 104
AY040066.1 - 2 - 3 201 104
AY050407.1 - 2 - 3 282 104
AY050931.1 - 2 - 3 250 104
AY056385.1 - 2 - 3 280 104
AY302079.1 - 2 - 3 201 104
BT002729.1 - 2 - 3 201 104

View Full Product Details