Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CSLD4
- Entrez Gene ID
-
829975
- Gene Name
- cellulose synthase like D4
- Gene Aliases
-
ARABIDOPSIS THALIANA CELLULOSE SYNTHASE-LIKE D4, AT4G38190, ATCSLD4, F20D10.310, F20D10_310, cellulose synthase like D4
- Chromosome Location
-
Chr.4: 17909832 - 17913790 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119980.3 |
NP_195532.1 |
2 - 3 |
2172 |
150 |
GenBank mRNA |
KJ138716.1 |
- |
2 - 3 |
2023 |
150 |
View Full Product Details