Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
URED
Entrez Gene ID
818068
Gene Name
urease accessory protein D
Gene Aliases
AT2G35035, urease accessory protein D
Chromosome Location
Chr.2: 14763178 - 14765079 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202751.1 NP_001189680.1 7 - 8 514 79
NM_179908.3 NP_850239.1 6 - 7 603 79
GenBank mRNA AJ312202.1 - 6 - 7 603 79

View Full Product Details