Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARAD1
Entrez Gene ID
818076
Gene Name
Exostosin family protein
Gene Aliases
ARABINAN DEFICIENT 1, ARABINOSYLTRANSFERASE, AT2G35100, T4C15.23, T4C15_23
Chromosome Location
Chr.2: 14793567 - 14795701 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179910.3 NP_850241.1 1 - 2 929 143
GenBank mRNA AK228986.1 - 1 - 2 809 143
BT015054.1 - 1 - 2 839 143
BT015854.1 - 1 - 2 792 143
KJ138874.1 - 1 - 2 792 143

View Full Product Details