Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GSL8
- Entrez Gene ID
-
818258
- Gene Name
- glucan synthase-like 8
- Gene Aliases
-
AT2G36850, ATGSL08, ATGSL8, CHOR, CHORUS, ENLARGED TETRAD 2, ET2, GLUCAN SYNTHASE-LIKE 8, GSL08, MAS, MASSUE, T1J8.3, T1J8_3, glucan synthase-like 8
- Chromosome Location
-
Chr.2: 15454671 - 15469964 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_179940.6 |
NP_850271.5 |
8 - 9 |
1315 |
99 |
GenBank mRNA |
GQ373182.1 |
- |
8 - 9 |
1017 |
99 |
View Full Product Details