Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AN
- Entrez Gene ID
-
839401
- Gene Name
- NAD(P)-binding Rossmann-fold superfamily protein
- Gene Aliases
-
ANGUSTIFOLIA, AT1G01510, DETORQUEO, DOQ, F22L4.6, F22L4_6
- Chromosome Location
-
Chr.1: 187145 - 190472 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100033.3 |
NP_563629.1 |
5 - 6 |
1813 |
78 |
View Full Product Details