Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATAF1
Entrez Gene ID
839265
Gene Name
NAC (No Apical Meristem) domain transcriptional regulator superfamily protein
Gene Aliases
ANAC002, ANAC2, AT1G01720, Arabidopsis NAC domain containing protein 2, T1N6.12, T1N6_12
Chromosome Location
Chr.1: 267993 - 269819 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100054.3 NP_171677.1 1 - 2 638 101
GenBank mRNA AF370617.1 - 1 - 2 160 101
BT024513.1 - 1 - 2 160 101

View Full Product Details