Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARK2
Entrez Gene ID
839306
Gene Name
armadillo repeat kinesin 2
Gene Aliases
AT1G01950, Arabidopsis thaliana KINESIN Ungrouped clade, AtKINUb, F22M8.8, F22M8_8, armadillo repeat kinesin 2, gene B
Chromosome Location
Chr.1: 325316 - 330619 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160828.1 NP_001154300.1 11 - 12 1730 117
NM_001160829.1 NP_001154301.1 11 - 12 1730 117
NM_001331317.1 NP_001320318.1 10 - 11 1872 117
NM_100075.8 NP_171697.3 11 - 12 1793 117
GenBank mRNA AB290929.1 - 11 - 12 1642 117

View Full Product Details