The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160828.1 | NP_001154300.1 | 15 - 16 | 2165 | 90 |
NM_001160829.1 | NP_001154301.1 | 16 - 17 | 2279 | 90 | |
NM_001331317.1 | NP_001320318.1 | 14 - 15 | 2358 | 90 | |
NM_100075.8 | NP_171697.3 | 15 - 16 | 2279 | 90 | |
GenBank mRNA | AB290929.1 | - | 15 - 16 | 2128 | 90 |