Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EDA10
- Entrez Gene ID
-
839301
- Gene Name
- SEC7-like guanine nucleotide exchange family protein
- Gene Aliases
-
AT1G01960, F22M8.9, F22M8_9, embryo sac development arrest 10
- Chromosome Location
-
Chr.1: 330588 - 337912 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100076.3 |
NP_171698.1 |
7 - 8 |
3653 |
69 |
View Full Product Details