Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG18G
Entrez Gene ID
838702
Gene Name
autophagy 18 G-like protein
Gene Aliases
AT1G03380, ATATG18G, F21B7.1, homolog of yeast autophagy 18 (ATG18) G
Chromosome Location
Chr.1: 835767 - 841373 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100220.8 NP_171837.7 3 - 4 1464 137
GenBank mRNA BT002410.1 - 3 - 4 1405 137

View Full Product Details