Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SOM
- Entrez Gene ID
-
839408
- Gene Name
- Zinc finger C-x8-C-x5-C-x3-H type family protein
- Gene Aliases
-
AT1G03790, AtTZF4, F21M11.30, F21M11_30, SOMNUS, TZF4, Tandem CCCH Zinc Finger protein 4
- Chromosome Location
-
Chr.1: 954290 - 956144 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100258.3 |
NP_171875.1 |
1 - 1 |
1579 |
110 |
View Full Product Details