Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCC5
Entrez Gene ID
839277
Gene Name
multidrug resistance-associated protein 5
Gene Aliases
ARABIDOPSIS THALIANA ATP BINDING CASSETTE TRANSPORTER C5, AT1G04120, ATABCC5, ATMRP5, ATP BINDING CASSETTE TRANSPORTER C5, ATP-binding cassette C5, Arabidopsis thaliana ATP-binding cassette C5, MULTIDRUG RESISTANCE PROTEIN 5, multidrug resistance-associated protein 5
Chromosome Location
Chr.1: 1064264 - 1071080 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197977.1 NP_001184906.1 3 - 4 2814 81
NM_001331463.1 NP_001322077.1 4 - 5 2681 81
NM_100293.3 NP_171908.1 3 - 4 2847 81
GenBank mRNA AF225908.1 - 3 - 4 2582 81
Y11250.1 - - 2562 81

View Full Product Details