Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AO4
Entrez Gene ID
839488
Gene Name
aldehyde oxidase 4
Gene Aliases
AAO4, ALDEHYDE OXIDASE 2, ALDEHYDE OXIDASE 4, AT1G04580, ATAO-4, ATAO2, T1G11.17, T1G11_17, aldehyde oxidase 4, arabdopsis thaliana aldehyde oxidase 4
Chromosome Location
Chr.1: 1252005 - 1257893 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100337.3 NP_563711.1 6 - 7 3786 66
GenBank mRNA AB037271.1 - 6 - 7 3403 66

View Full Product Details