Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAB1
Entrez Gene ID
839450
Gene Name
potassium channel beta subunit 1
Gene Aliases
AT1G04690, K+ CHANNEL BETA SUBUNIT PROTEIN, KV-BETA1, POTASSIUM CHANNEL BETA SUBUNIT HOMOLOG, T1G11.6, T1G11_6, potassium channel beta subunit 1
Chromosome Location
Chr.1: 1313443 - 1315823 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100349.4 NP_171963.1 3 - 4 926 66
GenBank mRNA AF061570.1 - 3 - 4 790 66
AY050821.1 - 3 - 4 760 66
AY091424.1 - 3 - 4 707 66

View Full Product Details